Product Details

SNP ID
rs2668758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47159308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGTCACCATTGCATATGCAATTG[A/C]CCAAAGAGCCAAGATAACAAGAAAT
Phenotype
MIM: 609382
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HDHD2 PubMed Links

Gene Details

Gene
HDHD2
Gene Name
haloacid dehalogenase like hydrolase domain containing 2
There are no transcripts associated with this gene.

Gene
IER3IP1
Gene Name
immediate early response 3 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016097.4 Intron NP_057181.1

View Full Product Details