Product Details

SNP ID
rs11828101
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:783045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGACCCGTGGACACCTCTGTCCC[C/T]AGAGATGATGCTGCGCGTTGGAGCT
Phenotype
MIM: 608213 MIM: 609302
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CEND1 PubMed Links
Additional Information
For this assay, SNP(s) [rs78797428] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CEND1
Gene Name
cell cycle exit and neuronal differentiation 1
There are no transcripts associated with this gene.

Gene
LOC171391
Gene Name
uncharacterized LOC171391
There are no transcripts associated with this gene.

Gene
PDDC1
Gene Name
Parkinson disease 7 domain containing 1
There are no transcripts associated with this gene.

Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
There are no transcripts associated with this gene.

View Full Product Details