Product Details
- SNP ID
-
rs11636728
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99581527 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAACTCTTTCTTTTTATTGACCTCC[C/T]GTGTTTTTCTTTCAACTGGTGAATA
- Phenotype
-
MIM: 600660
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MEF2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117912280] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MEF2A
- Gene Name
- myocyte enhancer factor 2A
View Full Product Details