Product Details
- SNP ID
-
rs2369476
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
62
- Location
-
Chr.1:160151698 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTCATCACTGAGGCACCTGGTTAC[G/T]CTTCACCTCTTGTTTCCTGCCCTCA
- Phenotype
-
MIM: 182340
MIM: 607321
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP1A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74123261] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP1A2
- Gene Name
- ATPase Na+/K+ transporting subunit alpha 2
There are no transcripts associated with this gene.
- Gene
- ATP1A4
- Gene Name
- ATPase Na+/K+ transporting subunit alpha 4
View Full Product Details