Product Details
- SNP ID
-
rs12625395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:26202054 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAACGTGACTCAATTAGAGGTCA[A/G]TTACTCCTTAGCAATGTATCAGAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR663A
PubMed Links
Gene Details
- Gene
- MIR663A
- Gene Name
- microRNA 663a
There are no transcripts associated with this gene.
- Gene
- MIR663AHG
- Gene Name
- MIR663A host gene
There are no transcripts associated with this gene.
View Full Product Details