Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012759.2 | 58 | UTR 5 | NP_001012777.1 | ||
NM_001012762.2 | 58 | UTR 5 | NP_001012780.1 | ||
NM_001318507.1 | 58 | UTR 5 | NP_001305436.1 | ||
NM_001318513.1 | 58 | UTR 5 | NP_001305442.1 | ||
XM_017023210.1 | 58 | Intron | XP_016878699.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171815.1 | 58 | Intron | NP_001165286.1 | ||
NM_001171816.1 | 58 | Intron | NP_001165287.1 | ||
NM_178841.3 | 58 | Intron | NP_849163.1 | ||
XM_011522845.2 | 58 | Intron | XP_011521147.1 | ||
XM_011522846.1 | 58 | Intron | XP_011521148.1 | ||
XM_011522847.1 | 58 | Intron | XP_011521149.1 | ||
XM_017022910.1 | 58 | Intron | XP_016878399.1 | ||
XM_017022911.1 | 58 | Intron | XP_016878400.1 | ||
XM_017022912.1 | 58 | Intron | XP_016878401.1 | ||
XM_017022913.1 | 58 | Intron | XP_016878402.1 |