Product Details

SNP ID
rs898010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5806675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCGAGGTTCATAGCTCCGGCACC[A/G]ACTCCACCCCCAGGGAGCTACAGAA
Phenotype
MIM: 602018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRTN PubMed Links

Gene Details

Gene
NRTN
Gene Name
neurturin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528041.2 Intron XP_011526343.1

View Full Product Details