Product Details

SNP ID
rs857979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37625791 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGCACACAAGTTTTCGCATGTGC[A/G]TGGCTATGTCTGGTCCACTCTGCTC
Phenotype
MIM: 600877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ6 PubMed Links

Gene Details

Gene
KCNJ6
Gene Name
potassium voltage-gated channel subfamily J member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002240.4 Intron NP_002231.1

View Full Product Details