Product Details

SNP ID
rs3741691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71664636 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGGAGTTCCTATTGTGGCTATC[G/T]CTTGTGTGGAAGGAACAGGAGGATT
Phenotype
MIM: 612531
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
THAP2 PubMed Links

Gene Details

Gene
THAP2
Gene Name
THAP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031435.3 Intron NP_113623.1
Gene
ZFC3H1
Gene Name
zinc finger C3H1-type containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144982.4 Intron NP_659419.3

View Full Product Details