Product Details

SNP ID
rs7305505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71613304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAAGAGCCTATTCCATTAGGCA[A/G]AGGACCAAAAGAATGTTAAGTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFC3H1 PubMed Links

Gene Details

Gene
ZFC3H1
Gene Name
zinc finger C3H1-type containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144982.4 Intron NP_659419.3

View Full Product Details