Product Details

SNP ID
rs1044605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65320605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAAGACCAAAGTGCACTGCTGC[C/T]CACACAGTTCCTACCTTTCTGGCTT
Phenotype
MIM: 606132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP2 PubMed Links

Gene Details

Gene
CDC42EP2
Gene Name
CDC42 effector protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006779.3 310 UTR 5 NP_006770.1
XM_017017094.1 310 UTR 5 XP_016872583.1

View Full Product Details