Product Details

SNP ID
rs13897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66563698 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCCAAGATACCACCCTTCACCC[C/T]GACATCCTCCTAAGCTACCACAATT
Phenotype
MIM: 102574 MIM: 603539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104.3 1530 Intron NP_001095.2
NM_001258371.2 1530 Intron NP_001245300.2
Gene
CTSF
Gene Name
cathepsin F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003793.3 1530 UTR 3 NP_003784.2
XM_011545328.2 1530 UTR 3 XP_011543630.1

View Full Product Details