Product Details

SNP ID
rs812265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:81474301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAAAATAAAACTAGAAGCAAAA[G/T]GAATAGTTTCAAGACAGTCGGCTAA
Phenotype
MIM: 602329
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEL1L PubMed Links

Gene Details

Gene
SEL1L
Gene Name
SEL1L ERAD E3 ligase adaptor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244984.1 5173 Intron NP_001231913.1
NM_005065.5 5173 UTR 3 NP_005056.3
XM_005267988.2 5173 UTR 3 XP_005268045.1
XM_005267989.3 5173 UTR 3 XP_005268046.1

View Full Product Details