Product Details

SNP ID
rs826263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36928934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTGTCTTTAACCTACATAGATT[G/T]ATCAGAATCTCAATAACAATTCTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF568 PubMed Links

Gene Details

Gene
ZNF568
Gene Name
zinc finger protein 568
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204835.1 Intron NP_001191764.1
NM_001204836.1 Intron NP_001191765.1
NM_001204837.1 Intron NP_001191766.1
NM_001204838.1 Intron NP_001191767.1
NM_001204839.1 Intron NP_001191768.1
NM_198539.3 Intron NP_940941.2
XM_017026772.1 Intron XP_016882261.1
XM_017026773.1 Intron XP_016882262.1
XM_017026774.1 Intron XP_016882263.1
XM_017026775.1 Intron XP_016882264.1
XM_017026776.1 Intron XP_016882265.1
XM_017026777.1 Intron XP_016882266.1
XM_017026778.1 Intron XP_016882267.1
XM_017026779.1 Intron XP_016882268.1
XM_017026780.1 Intron XP_016882269.1

View Full Product Details