Product Details

SNP ID
rs851609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42232115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGGACCTCAAGTGAGGAGGGAT[A/G]AGAATGGCGTTGAGAGACGGTTGGA
Phenotype
MIM: 606784 MIM: 614387
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSK3A PubMed Links

Gene Details

Gene
GSK3A
Gene Name
glycogen synthase kinase 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019884.2 1439 Silent Mutation CTC,CTT L440L NP_063937.2
Gene
ZNF526
Gene Name
zinc finger protein 526
There are no transcripts associated with this gene.

View Full Product Details