Product Details
- SNP ID
-
rs854484
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:201286061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTATCTGGCTTAGTTTTCATCAGA[A/T]TTCCTTGTCTTCTCCATCAGAAACA
- Phenotype
-
MIM: 601975
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PKP1
PubMed Links
Gene Details
- Gene
- PKP1
- Gene Name
- plakophilin 1
View Full Product Details