Product Details

SNP ID
rs1053401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:426875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCAGCTTGAACATCTTCTTCA[G/T]GCTCTTCTGGATTGGTATGAGTTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCA11P PubMed Links

Gene Details

Gene
ABCA11P
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
There are no transcripts associated with this gene.

View Full Product Details