Product Details

SNP ID
rs903517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73471899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTTGTGCACCCATCACCACCAA[C/T]GGTCTCTTGAACTTTTTCATCATCC
Phenotype
MIM: 614667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTO1 PubMed Links

Gene Details

Gene
MTO1
Gene Name
mitochondrial tRNA translation optimization 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123226.1 Intron NP_001116698.1
NM_012123.3 Intron NP_036255.2
NM_133645.2 Intron NP_598400.1
XM_006715444.2 Intron XP_006715507.1
XM_006715445.2 Intron XP_006715508.1
XM_006715446.3 Intron XP_006715509.1
XM_011535723.1 Intron XP_011534025.1
XM_011535724.1 Intron XP_011534026.1
XM_011535725.2 Intron XP_011534027.1
XM_017010697.1 Intron XP_016866186.1
XM_017010698.1 Intron XP_016866187.1
XM_017010699.1 Intron XP_016866188.1
XM_017010700.1 Intron XP_016866189.1

View Full Product Details