Product Details

SNP ID
rs1472694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19121526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAACTGTGTCTTATTCATCTTT[C/T]ATCCCTGATTACCTACTTCTGTGAT
Phenotype
MIM: 612815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC13 PubMed Links

Gene Details

Gene
ZDHHC13
Gene Name
zinc finger DHHC-type containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001483.2 Intron NP_001001483.1
NM_019028.2 Intron NP_061901.2
XM_005252996.2 Intron XP_005253053.1
XM_011520193.1 Intron XP_011518495.1
XM_011520194.2 Intron XP_011518496.1
XM_011520195.1 Intron XP_011518497.1

View Full Product Details