Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142289.2 | Intron | NP_001135761.2 | |||
NM_001142290.2 | Intron | NP_001135762.1 | |||
NM_001142291.2 | Intron | NP_001135763.2 | |||
NM_015246.3 | Intron | NP_056061.1 | |||
XM_005255217.1 | Intron | XP_005255274.1 | |||
XM_005255218.1 | Intron | XP_005255275.1 | |||
XM_005255219.3 | Intron | XP_005255276.1 | |||
XM_005255220.3 | Intron | XP_005255277.1 | |||
XM_005255221.3 | Intron | XP_005255278.1 | |||
XM_011522441.1 | Intron | XP_011520743.1 | |||
XM_011522442.2 | Intron | XP_011520744.1 | |||
XM_017023091.1 | Intron | XP_016878580.1 | |||
XM_017023092.1 | Intron | XP_016878581.1 | |||
XM_017023093.1 | Intron | XP_016878582.1 |