Product Details

SNP ID
rs1641107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8678720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTCCATGGCAGAGCTTGCAACAC[C/T]GGTATGGGAAGGGATTACAGTATAT
Phenotype
MIM: 137150 MIM: 615261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABAT PubMed Links

Gene Details

Gene
ABAT
Gene Name
4-aminobutyrate aminotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000663.4 Intron NP_000654.2
NM_001127448.1 Intron NP_001120920.1
NM_020686.5 Intron NP_065737.2
XM_011522400.2 Intron XP_011520702.1
XM_011522401.2 Intron XP_011520703.1
Gene
METTL22
Gene Name
methyltransferase like 22
There are no transcripts associated with this gene.

View Full Product Details