Product Details

SNP ID
rs9547190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36302566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCTGACCTCTGGTCAAAGCTAA[C/T]TGTTTAAAGTTTATGTTCCATTATT
Phenotype
MIM: 607111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC169 PubMed Links

Gene Details

Gene
CCDC169
Gene Name
coiled-coil domain containing 169
There are no transcripts associated with this gene.

Gene
CCDC169-SOHLH2
Gene Name
CCDC169-SOHLH2 readthrough
There are no transcripts associated with this gene.

Gene
SPG20
Gene Name
spastic paraplegia 20 (Troyer syndrome)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142294.1 4087 UTR 3 NP_001135766.1
NM_001142295.1 4087 UTR 3 NP_001135767.1
NM_001142296.1 4087 UTR 3 NP_001135768.1
NM_015087.4 4087 UTR 3 NP_055902.1
XM_005266313.4 4087 UTR 3 XP_005266370.1
XM_005266314.3 4087 UTR 3 XP_005266371.1
XM_005266315.3 4087 UTR 3 XP_005266372.1
XM_005266317.3 4087 UTR 3 XP_005266374.1
XM_011535012.2 4087 UTR 3 XP_011533314.1

View Full Product Details