Product Details

SNP ID
rs3753753
Assay Type
Validated
NCBI dbSNP Submissions
48
Location
Chr.1:52603842 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TGCTTTGCACCTGTGTTTGGTGGAT[C/G]CCAGCAGGAACTTCATTTAGAACTC
Phenotype
MIM: 615784
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPX7 PubMed Links

Gene Details

Gene
GPX7
Gene Name
glutathione peroxidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015696.4 Intron NP_056511.2

View Full Product Details