Product Details

SNP ID
rs2650757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21520743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAATTATCTGATATTTGCAGTCC[A/G]AAGTACTTACACAGAAAAAGCAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF429 PubMed Links

Gene Details

Gene
ZNF429
Gene Name
zinc finger protein 429
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001415.2 Intron NP_001001415.2
XM_017026748.1 Intron XP_016882237.1
XM_017026749.1 Intron XP_016882238.1
XM_017026750.1 Intron XP_016882239.1
XM_017026751.1 Intron XP_016882240.1
XM_017026752.1 Intron XP_016882241.1
XM_017026753.1 Intron XP_016882242.1
XM_017026754.1 Intron XP_016882243.1
XM_017026755.1 Intron XP_016882244.1
XM_017026756.1 Intron XP_016882245.1
XM_017026757.1 Intron XP_016882246.1

View Full Product Details