Product Details

SNP ID
rs11767129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73861439 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGCCCCACTCCAAACTACAGTG[C/T]TGACATTTTCTTTTCTTTTCTTTAT
Phenotype
MIM: 612547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WBSCR28 PubMed Links

Gene Details

Gene
WBSCR28
Gene Name
Williams-Beuren syndrome chromosome region 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182504.3 Intron NP_872310.2
XM_011515785.2 Intron XP_011514087.1
XM_017011741.1 Intron XP_016867230.1

View Full Product Details