Product Details

SNP ID
rs28700162
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:18872979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTGCGGAAAAGACACTTCTGT[A/T]TTTCTCTGAAAAGATGCAACAGCCT
Phenotype
MIM: 606811
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH4A1 PubMed Links

Gene Details

Gene
ALDH4A1
Gene Name
aldehyde dehydrogenase 4 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161504.1 Intron NP_001154976.1
NM_001319218.1 Intron NP_001306147.1
NM_003748.3 Intron NP_003739.2
NM_170726.2 Intron NP_733844.1

View Full Product Details