Product Details

SNP ID
rs4283241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29977028 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGATAATGCACGTAAGGTACTCA[A/G]TAAATGTTATTTGTACCCTGCAAAT
Phenotype
MIM: 613199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107987231 PubMed Links

Gene Details

Gene
LOC107987231
Gene Name
uncharacterized LOC107987231
There are no transcripts associated with this gene.

Gene
TAOK2
Gene Name
TAO kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252043.1 Intron NP_001238972.1
NM_004783.3 Intron NP_004774.1
NM_016151.3 Intron NP_057235.2
XM_011545982.1 Intron XP_011544284.1
XM_011545983.2 Intron XP_011544285.1
XM_011545984.1 Intron XP_011544286.1
XM_011545985.1 Intron XP_011544287.1
XM_011545986.2 Intron XP_011544288.1
Gene
TMEM219
Gene Name
transmembrane protein 219
There are no transcripts associated with this gene.

View Full Product Details