Product Details

SNP ID
rs4567786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75777029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGAGAAAGCAATTTTAATGGGG[C/T]ATAACTTTAAGCAGTTGCTAACTTC
Phenotype
MIM: 601058 MIM: 616375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H3F3B PubMed Links

Gene Details

Gene
H3F3B
Gene Name
H3 histone, family 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005324.4 2266 UTR 3 NP_005315.1
Gene
MIR4738
Gene Name
microRNA 4738
There are no transcripts associated with this gene.

Gene
UNK
Gene Name
unkempt family zinc finger
There are no transcripts associated with this gene.

View Full Product Details