Product Details

SNP ID
rs3935147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178119280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCCCACACAGGCCCCGAGGTGG[A/G]GCTGGCCCTTCGGTGCTCGGCTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
N4BP3 PubMed Links

Gene Details

Gene
N4BP3
Gene Name
NEDD4 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015111.1 Intron NP_055926.1
XM_006714834.3 Intron XP_006714897.1
XM_011534473.1 Intron XP_011532775.1
XM_011534474.1 Intron XP_011532776.1

View Full Product Details