Product Details

SNP ID
rs13138942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26865141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACTGTATTCTCTTTCTCTTTCC[C/T]CTTTAGTATCATTACCAGTGCTTTT
Phenotype
MIM: 610841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105374546 PubMed Links

Gene Details

Gene
LOC105374546
Gene Name
uncharacterized LOC105374546
There are no transcripts associated with this gene.

Gene
STIM2
Gene Name
stromal interaction molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169117.1 Intron NP_001162588.1
NM_001169118.1 Intron NP_001162589.1
NM_020860.3 Intron NP_065911.3
Gene
TBC1D19
Gene Name
TBC1 domain family member 19
There are no transcripts associated with this gene.

View Full Product Details