Product Details

SNP ID
rs6823775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15606234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAGCAATTACAATAAATATTAT[A/C]TTCAAATTTTAAAGCAAAATAAACT
Phenotype
MIM: 612013 MIM: 605655
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CC2D2A PubMed Links

Gene Details

Gene
CC2D2A
Gene Name
coiled-coil and C2 domain containing 2A
There are no transcripts associated with this gene.

Gene
FBXL5
Gene Name
F-box and leucine rich repeat protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193534.1 Intron NP_001180463.1
NM_001193535.1 Intron NP_001180464.1
NM_012161.3 Intron NP_036293.1
XM_006713959.3 Intron XP_006714022.1
XM_011513831.2 Intron XP_011512133.1
XM_011513832.2 Intron XP_011512134.1
XM_011513833.2 Intron XP_011512135.1
XM_017008019.1 Intron XP_016863508.1

View Full Product Details