Product Details

SNP ID
rs35465650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131833029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACACAGCACAGTGGTCACAGGC[A/G]GAGACCCGGAAGCCAACAGCCTGGG
Phenotype
MIM: 602285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP17 PubMed Links

Gene Details

Gene
MMP17
Gene Name
matrix metallopeptidase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016155.5 Intron NP_057239.4
XM_011538355.2 Intron XP_011536657.1
XM_011538356.2 Intron XP_011536658.1
XM_011538357.2 Intron XP_011536659.1
XM_017019307.1 Intron XP_016874796.1

View Full Product Details