Product Details
- SNP ID
-
rs3738722
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:226633236 on Build GRCh38
- Set Membership
-
JSNP
Validated
- Context Sequence [VIC/FAM]
- CAGGTTGACAGTTCCCCAGAGACCC[T/C]GGGGTGGTGGGGTGGGCAGTGGTTC
- Phenotype
-
MIM: 147522
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITPKB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144827491,rs149442069] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITPKB
- Gene Name
- inositol-trisphosphate 3-kinase B
View Full Product Details