Product Details

SNP ID
rs3116466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103300463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTTATGTTTTGTGTACCTTTG[C/T]GGCCCTGTTTAGTAGATTCGTTTGA
Phenotype
MIM: 603409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FZD6 PubMed Links

Gene Details

Gene
FZD6
Gene Name
frizzled class receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164615.1 Intron NP_001158087.1
NM_001164616.1 Intron NP_001158088.1
NM_001317796.1 Intron NP_001304725.1
NM_003506.3 Intron NP_003497.2

View Full Product Details