Product Details

SNP ID
rs9655407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39952074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGCGAAGGGAACTTTTCTCCTA[A/G]CTGTGGCAGGGAAGATTTCTAAGGA
Phenotype
MIM: 603309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK13 PubMed Links

Gene Details

Gene
CDK13
Gene Name
cyclin dependent kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003718.4 Intron NP_003709.3
NM_031267.3 Intron NP_112557.2
XM_011515597.2 Intron XP_011513899.1
XM_017012750.1 Intron XP_016868239.1
XM_017012751.1 Intron XP_016868240.1

View Full Product Details