Product Details

SNP ID
rs13382200
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:188296286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGGGATATGAAATTGAAATCAG[C/G]CCTTTATTTTTAAAGAGAATGGCTA
Phenotype
MIM: 608165
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GULP1 PubMed Links

Gene Details

Gene
GULP1
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252668.1 Intron NP_001239597.1
NM_001252669.1 Intron NP_001239598.1
NM_016315.3 Intron NP_057399.1
XM_006712580.3 Intron XP_006712643.1
XM_006712581.1 Intron XP_006712644.1
XM_006712582.1 Intron XP_006712645.1
XM_006712583.3 Intron XP_006712646.1
XM_006712584.3 Intron XP_006712647.1
XM_006712585.1 Intron XP_006712648.1
XM_006712589.3 Intron XP_006712652.1
XM_011511329.1 Intron XP_011509631.1
XM_011511331.1 Intron XP_011509633.1
XM_011511332.1 Intron XP_011509634.1
XM_011511333.1 Intron XP_011509635.1
XM_011511335.2 Intron XP_011509637.1
XM_017004303.1 Intron XP_016859792.1
XM_017004304.1 Intron XP_016859793.1
XM_017004305.1 Intron XP_016859794.1
XM_017004306.1 Intron XP_016859795.1
XM_017004307.1 Intron XP_016859796.1
XM_017004308.1 Intron XP_016859797.1
Gene
LINC01090
Gene Name
long intergenic non-protein coding RNA 1090
There are no transcripts associated with this gene.

Gene
MIR561
Gene Name
microRNA 561
There are no transcripts associated with this gene.

View Full Product Details