Product Details
- SNP ID
-
hCV309084
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:38773268 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTGGCTAATACATTAACATTAA[T/C]AGCAGCTCGAAGGTTTGCCCAGAAA
- Phenotype
-
MIM: 606270
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLR10
PubMed Links
Gene Details
- Gene
- TLR10
- Gene Name
- toll like receptor 10
There are no transcripts associated with this gene.
View Full Product Details