Product Details

SNP ID
rs10776483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38773419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTTCCAGTAAGATAAGAATTAT[A/G]TGATCAGAATTTTCATGGAAGAGAT
Phenotype
MIM: 606270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR10 PubMed Links

Gene Details

Gene
TLR10
Gene Name
toll like receptor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017388.2 2304 Silent Mutation CAC,CAT H724H NP_001017388.1
NM_001195106.1 2304 Silent Mutation CAC,CAT H724H NP_001182035.1
NM_001195107.1 2304 Silent Mutation CAC,CAT H724H NP_001182036.1
NM_001195108.1 2304 Silent Mutation CAC,CAT H710H NP_001182037.1
NM_030956.3 2304 Silent Mutation CAC,CAT H724H NP_112218.2
XM_011513760.2 2304 Silent Mutation CAC,CAT H710H XP_011512062.1
XM_011513761.2 2304 Silent Mutation CAC,CAT H724H XP_011512063.1
XM_011513762.2 2304 Silent Mutation CAC,CAT H724H XP_011512064.1

View Full Product Details