Product Details
- SNP ID
-
rs9889352
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:32448743 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTCGAAACAACTTGAAGTAGGTT[C/T]TGTTACCATTTTACACATGAGGATA
- Phenotype
-
MIM: 604449
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PSMD11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112159436] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSMD11
- Gene Name
- proteasome 26S subunit, non-ATPase 11
View Full Product Details