Product Details
- SNP ID
-
rs3793451
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:69044364 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGTTATCATGGCTGCCCTCTCACTT[C/T]TTCAGAGACATGTGTTTCTAAGGTC
- Phenotype
-
MIM: 606829
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FXN
PubMed Links
Gene Details
- Gene
- FXN
- Gene Name
- frataxin
View Full Product Details