Product Details

SNP ID
hCV326811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37341421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACCTGCCTTTCCTCTCCAGCT[A/C]TGGGCCAAGCCCACTGTGCTCACCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELFN2 PubMed Links

Gene Details

Gene
ELFN2
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052906.4 Intron NP_443138.2

View Full Product Details