Product Details

SNP ID
rs11962777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132769170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGTCTGATATTTATTTATTGTT[C/T]GTCTCTTCCACCAGTATAAGTTACA
Phenotype
MIM: 613361 MIM: 603571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC18B1 PubMed Links

Gene Details

Gene
SLC18B1
Gene Name
solute carrier family 18 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052831.2 Intron NP_439896.1
XM_017010236.1 Intron XP_016865725.1
Gene
VNN2
Gene Name
vanin 2
There are no transcripts associated with this gene.

View Full Product Details