Product Details
- SNP ID
-
rs2282810
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150173136 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACCCTCCCATGGCTTCCTTTTGTT[C/T]TTGAAACCAAATCCATATCCCTCTC
- Phenotype
-
MIM: 600746
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDX1
PubMed Links
Gene Details
- Gene
- CDX1
- Gene Name
- caudal type homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001804.2 |
|
Intron |
|
|
NP_001795.2 |
View Full Product Details