Product Details

SNP ID
rs12503909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17579675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGCACATGTAGCTGACCCCAG[C/T]ACCTGCTTCTTTTTTTGCTTTCTTC
Phenotype
MIM: 170250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAP3 PubMed Links

Gene Details

Gene
LAP3
Gene Name
leucine aminopeptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015907.2 Intron NP_056991.2

View Full Product Details