Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003833.4 | 80 | UTR 5 | NP_003824.2 | ||
NM_030590.3 | 80 | Intron | NP_085080.1 | ||
NM_030592.3 | 80 | Intron | NP_085095.1 | ||
XM_005260597.1 | 80 | UTR 5 | XP_005260654.1 | ||
XM_017028113.1 | 80 | UTR 5 | XP_016883602.1 | ||
XM_017028114.1 | 80 | UTR 5 | XP_016883603.1 | ||
XM_017028115.1 | 80 | UTR 5 | XP_016883604.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281448.1 | 80 | Intron | NP_001268377.1 | ||
NM_001281449.1 | 80 | Intron | NP_001268378.1 | ||
NM_014276.3 | 80 | Intron | NP_055091.2 | ||
XM_011528521.2 | 80 | Intron | XP_011526823.1 | ||
XM_011528522.2 | 80 | Intron | XP_011526824.1 | ||
XM_011528523.2 | 80 | Intron | XP_011526825.1 |