Product Details
- SNP ID
-
rs11845657
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36658359 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGGACAAGAGGCGGGCACCGTC[G/T]CCTGGGCCTCGCTTGGGGGGGGGGG
- Phenotype
-
MIM: 167416
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PAX9
PubMed Links
Gene Details
- Gene
- PAX9
- Gene Name
- paired box 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006194.3 |
|
Intron |
|
|
NP_006185.1 |
View Full Product Details