Product Details

SNP ID
rs6656720
Assay Type
Functionally Tested
NCBI dbSNP Submissions
29
Location
Chr.1:28506185 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGCTTCCCTAGGAAGATCATATA[A/G]TATTTTATTTATTTACTTTTTTTTT
Phenotype
MIM: 608726 MIM: 179710 MIM: 603238 MIM: 180645 MIM: 603239
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PHACTR4 PubMed Links
Additional Information
For this assay, SNP(s) [rs6696931] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PHACTR4
Gene Name
phosphatase and actin regulator 4
There are no transcripts associated with this gene.

Gene
RCC1
Gene Name
regulator of chromosome condensation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048194.2 Intron NP_001041659.1
NM_001048195.2 Intron NP_001041660.1
NM_001048199.2 Intron NP_001041664.1
NM_001269.4 Intron NP_001260.1
Gene
SNHG3
Gene Name
small nucleolar RNA host gene 3
There are no transcripts associated with this gene.

Gene
SNORA73A
Gene Name
small nucleolar RNA, H/ACA box 73A
There are no transcripts associated with this gene.

Gene
SNORA73B
Gene Name
small nucleolar RNA, H/ACA box 73B
There are no transcripts associated with this gene.

View Full Product Details