Product Details

SNP ID
rs4239134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15980653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCATGTGAAGGTCAAGTGTGG[C/T]GTCAGGCTGTCTCACAGGTAGTGCA
Phenotype
MIM: 600446 MIM: 614535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
There are no transcripts associated with this gene.

Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 Intron NP_001036162.1
NM_001042698.1 Intron NP_001036163.1

View Full Product Details