Product Details

SNP ID
rs12724106
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:197158448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTATCAGCCAAGTAAATGAAATC[A/G]AAAGTAGCATGTTTAATTAAATTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB41 PubMed Links

Gene Details

Gene
ZBTB41
Gene Name
zinc finger and BTB domain containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194314.2 3710 UTR 3 NP_919290.2
XM_011509507.2 3710 Intron XP_011507809.1
XM_017001205.1 3710 Intron XP_016856694.1

View Full Product Details