Product Details

SNP ID
rs16855690
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:205508398 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAACAGCTCAGGCCAAGGGTCTG[A/G]GATCCAGGATCACTACAGAGCTGGT
Phenotype
MIM: 169190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK18 PubMed Links

Gene Details

Gene
CDK18
Gene Name
cyclin dependent kinase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002596.3 Intron NP_002587.2
NM_212502.2 Intron NP_997667.1
NM_212503.2 Intron NP_997668.1
XM_011509602.2 Intron XP_011507904.1
XM_017001423.1 Intron XP_016856912.1

View Full Product Details